Assay Details
| Gene Symbol: PLVAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319556.1 | NP_001306485.1 | 3 - 4 | 1177 | 56 | |
| XM_005588380.2 | XP_005588437.1 | 3 - 4 | 1249 | 56 | ||
| GenBank mRNA | KM282622.1 | - | 3 - 4 | 1177 | 56 | |
More Information
Set Membership: |
|

PLVAP
Genomic Map