Assay Details
| Gene Symbol: MGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285149.1 | NP_001272078.1 | 1217 | 110 | ||
| GenBank mRNA | AB179178.1 | - | 1217 | 110 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGAT2| Gene Symbol: MGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285149.1 | NP_001272078.1 | 1217 | 110 | ||
| GenBank mRNA | AB179178.1 | - | 1217 | 110 | ||
Set Membership: |
|