Assay Details
Gene Symbol: ANAPC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578950.2 | XP_005579007.1 | 4 - 5 | 469 | 96 | |
XM_015435437.1 | XP_015290923.1 | 5 - 6 | 523 | 96 | ||
XM_015435438.1 | XP_015290924.1 | 5 - 6 | 401 | 96 | ||
XM_015435439.1 | XP_015290925.1 | 5 - 6 | 709 | 96 | ||
XM_015435440.1 | XP_015290926.1 | 4 - 5 | 464 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
