Assay Details
Gene Symbol: SMIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015442374.1 | XP_015297860.1 | 2 - 3 | 771 | 79 | |
XM_015442375.1 | XP_015297861.1 | 2 - 3 | 766 | 79 | ||
XM_015442376.1 | XP_015297862.1 | 2 - 3 | 767 | 79 | ||
XM_015442377.1 | XP_015297863.1 | 2 - 3 | 764 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
