Assay Details
| Gene Symbol: LOC102131997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590312.2 | XP_005590369.1 | 7 - 8 | 1622 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102131997| Gene Symbol: LOC102131997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590312.2 | XP_005590369.1 | 7 - 8 | 1622 | 63 | |
Set Membership: |
|