Assay Details
Gene Symbol: BIRC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015433410.1 | XP_015288896.1 | 1 - 2 | 45 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
