Assay Details
Gene Symbol: LOC102115187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015438037.1 | XP_015293523.1 | 1 | 115 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015438037.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
