Assay Details
Gene Symbol: RASL10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567674.1 | XP_005567731.1 | 1 - 2 | 305 | 103 | |
XM_015457325.1 | XP_015312811.1 | 1 - 2 | 305 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
