Assay Details
| Gene Symbol: FPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590156.2 | XP_005590213.1 | 1 - 2 | 1306 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005590156.2 transcripts.
Set Membership: |
|

FPR3
Genomic Map