Assay Details
| Gene Symbol: CEACAM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442086.1 | XP_015297572.1 | 3 - 4 | 818 | 84 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEACAM7| Gene Symbol: CEACAM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442086.1 | XP_015297572.1 | 3 - 4 | 818 | 84 | |
Set Membership: | 
		   
					
						
							 |