Assay Details
| Gene Symbol: LOC102137841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441692.1 | XP_015297178.1 | 1 - 2 | 860 | 81 | |
| XM_015441693.1 | XP_015297179.1 | 1 - 2 | 860 | 81 | ||
| XM_015441694.1 | XP_015297180.1 | 1 - 2 | 860 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015441692.1, XM_015441693.1, XM_015441694.1 transcripts.
Set Membership: |
|

LOC102137841
Genomic Map