Assay Details
| Gene Symbol: KHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449184.1 | XP_015304670.1 | 1 - 2 | 214 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

KHDC1| Gene Symbol: KHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449184.1 | XP_015304670.1 | 1 - 2 | 214 | 73 | |