Assay Details
Gene Symbol: TEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585016.2 | XP_005585073.1 | 3 - 4 | 551 | 72 | |
XM_015438870.1 | XP_015294356.1 | 3 - 4 | 280 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
