Assay Details
| Gene Symbol: SCP2D1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569662.1 | XP_005569719.1 | 3 - 4 | 115 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCP2D1-AS1| Gene Symbol: SCP2D1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569662.1 | XP_005569719.1 | 3 - 4 | 115 | 65 | |
Set Membership: |
|