Assay Details
| Gene Symbol: ZNF649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441341.1 | XP_015296827.1 | 2 - 3 | 309 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015441341.1 transcripts.
Set Membership: |
|

ZNF649
Genomic Map