Assay Details
| Gene Symbol: LOC107128534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443938.1 | XP_015299424.1 | 1 | 1615 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015443938.1 transcripts.
Set Membership: |
|

LOC107128534
Genomic Map