Assay Details
| Gene Symbol: LOC107126546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015437233.1 | XP_015292719.1 | 1 - 2 | 1575 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107126546| Gene Symbol: LOC107126546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015437233.1 | XP_015292719.1 | 1 - 2 | 1575 | 80 | |
Set Membership: |
|