Assay Details
Gene Symbol: LOC102135496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015453379.1 | XP_015308865.1 | 2 - 3 | 412 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
