Assay Details
| Gene Symbol: LOC102127474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015440431.1 | XP_015295917.1 | 3 - 4 | 375 | 87 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  3' Most  Recommended Assay  Probe spans exons | 
Panther Classification:
 Back To Top
 Back To Top
 LOC102127474
LOC102127474 Genomic Map
Genomic Map Back To Top
 Back To Top