Assay Details
Gene Symbol: CST9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568169.2 | XP_005568226.1 | 1 | 118 | 74 | |
XM_005568170.1 | XP_005568227.1 | 1 | 118 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005568169.2, XM_005568170.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
