Assay Details
Gene Symbol: LOC102135021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005586516.2 | XP_005586573.1 | 8 | 1221 | 108 | |
XM_005586517.2 | XP_005586574.1 | 8 | 1073 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
