Assay Details
Gene Symbol: GEMIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582413.2 | XP_005582470.1 | 2 | 2223 | 99 | |
XM_005582414.2 | XP_005582471.1 | 2 | 4055 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
