Assay Details
| Gene Symbol: Chrna7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007390.3 | NP_031416.3 | 1 - 2 | 110 | 102 | |
| GenBank mRNA | AF225980.1 | - | 1 - 2 | 100 | 102 | |
| AK034208.1 | - | 1 - 2 | 154 | 102 | ||
| AK157809.1 | - | 1 - 2 | 107 | 102 | ||
| L37663.1 | - | 1 - 2 | 110 | 102 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF225980.1,AK034208.1,AK157809.1,L37663.1,NM_007390.3 transcripts.
Set Membership: |
|

Chrna7
Genomic Map