Assay Details
Gene Symbol: Acvr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007396.4 | NP_031422.3 | 7 - 8 | 1625 | 90 | |
GenBank mRNA | AK041246.1 | - | 7 - 8 | 1578 | 90 | |
AK143186.1 | - | 8 - 9 | 1724 | 90 | ||
BC138823.1 | - | 7 - 8 | 1286 | 8861845 | 90 | |
M65287.1 | - | 7 - 8 | 1032 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
