Assay Details
| Gene Symbol: Adcyap1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025372.2 | NP_001020543.1 | 8 - 9 | 1007 | 61 | |
| NM_007407.4 | NP_031433.3 | 8 - 9 | 1007 | 61 | ||
| GenBank mRNA | AK042820.1 | - | 8 - 9 | 999 | 61 | |
| AK047304.1 | - | 7 - 8 | 510 | 61 | ||
| AK081121.1 | - | 7 - 8 | 510 | 61 | ||
| AK083108.1 | - | 8 - 9 | 1006 | 61 | ||
| AK147609.1 | - | 8 - 9 | 748 | 61 | ||
| BC067039.1 | - | 8 - 9 | 940 | 6852762 | 61 | |
| D82935.1 | - | 8 - 9 | 690 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adcyap1r1
Genomic Map