Assay Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007505.2 | NP_031531.1 | 7 - 8 | 1232 | 73 | |
| GenBank mRNA | AK043976.1 | - | 7 - 8 | 1000 | 73 | |
| AK076572.1 | - | 7 - 8 | 1000 | 73 | ||
| AK098155.1 | - | 7 - 8 | 1010 | 73 | ||
| AK146797.1 | - | 7 - 8 | 1003 | 73 | ||
| AK150426.1 | - | 7 - 8 | 1003 | 73 | ||
| AK150843.1 | - | 7 - 8 | 1000 | 73 | ||
| AK151004.1 | - | 7 - 8 | 1000 | 73 | ||
| AK151128.1 | - | 7 - 8 | 1009 | 73 | ||
| AK151224.1 | - | 7 - 8 | 1007 | 73 | ||
| AK151920.1 | - | 7 - 8 | 1004 | 73 | ||
| AK152054.1 | - | 7 - 8 | 1011 | 73 | ||
| AK152890.1 | - | 7 - 8 | 1003 | 73 | ||
| AK153167.1 | - | 7 - 8 | 1004 | 73 | ||
| AK153282.1 | - | 7 - 8 | 1000 | 73 | ||
| AK159491.1 | - | 7 - 8 | 1000 | 73 | ||
| AK159540.1 | - | 7 - 8 | 1000 | 73 | ||
| AK159758.1 | - | 7 - 8 | 1003 | 73 | ||
| AK160043.1 | - | 7 - 8 | 1010 | 73 | ||
| AK164110.1 | - | 7 - 8 | 1003 | 73 | ||
| AK164193.1 | - | 5 - 6 | 706 | 73 | ||
| AK166709.1 | - | 7 - 8 | 1003 | 73 | ||
| AK166812.1 | - | 7 - 8 | 1016 | 73 | ||
| AK167159.1 | - | 7 - 8 | 1000 | 73 | ||
| AK167863.1 | - | 7 - 8 | 980 | 73 | ||
| AK168198.1 | - | 7 - 8 | 1010 | 73 | ||
| AK168369.1 | - | 7 - 8 | 1002 | 73 | ||
| AK168617.1 | - | 7 - 8 | 1000 | 73 | ||
| AK168879.1 | - | 7 - 8 | 1000 | 73 | ||
| AK168890.1 | - | 7 - 8 | 1000 | 73 | ||
| AK168932.1 | - | 7 - 8 | 1000 | 73 | ||
| AK169080.1 | - | 7 - 8 | 1003 | 73 | ||
| AK169084.1 | - | 7 - 8 | 1003 | 73 | ||
| AK169105.1 | - | 7 - 8 | 1000 | 73 | ||
| AK169142.1 | - | 7 - 8 | 1045 | 73 | ||
| AK169300.1 | - | 7 - 8 | 1000 | 73 | ||
| AK169308.1 | - | 7 - 8 | 1005 | 73 | ||
| AK169414.1 | - | 7 - 8 | 1003 | 73 | ||
| BC014854.1 | - | 7 - 8 | 987 | 4500893 | 73 | |
| BC020417.1 | - | 7 - 8 | 987 | 73 | ||
| BC055892.1 | - | 7 - 8 | 1437 | 73 | ||
| L01062.1 | - | 7 - 8 | 987 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5a1
Genomic Map