Assay Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007505.2 | NP_031531.1 | 10 - 11 | 1705 | 85 | |
| GenBank mRNA | AK043976.1 | - | 10 - 11 | 1473 | 85 | |
| AK076572.1 | - | 10 - 11 | 1473 | 85 | ||
| AK098155.1 | - | 10 - 11 | 1483 | 85 | ||
| AK146797.1 | - | 10 - 11 | 1371 | 85 | ||
| AK150426.1 | - | 10 - 11 | 1476 | 85 | ||
| AK150843.1 | - | 10 - 11 | 1473 | 85 | ||
| AK151004.1 | - | 10 - 11 | 1473 | 85 | ||
| AK151128.1 | - | 10 - 11 | 1482 | 85 | ||
| AK151224.1 | - | 10 - 11 | 1480 | 85 | ||
| AK151920.1 | - | 10 - 11 | 1477 | 85 | ||
| AK152054.1 | - | 10 - 11 | 1484 | 85 | ||
| AK152890.1 | - | 10 - 11 | 1476 | 85 | ||
| AK159491.1 | - | 10 - 11 | 1473 | 85 | ||
| AK159540.1 | - | 10 - 11 | 1473 | 85 | ||
| AK159758.1 | - | 10 - 11 | 1476 | 85 | ||
| AK160043.1 | - | 10 - 11 | 1483 | 85 | ||
| AK164110.1 | - | 10 - 11 | 1476 | 85 | ||
| AK164193.1 | - | 8 - 9 | 1179 | 85 | ||
| AK166709.1 | - | 10 - 11 | 1476 | 85 | ||
| AK166812.1 | - | 10 - 11 | 1489 | 85 | ||
| AK167159.1 | - | 10 - 11 | 1473 | 85 | ||
| AK167863.1 | - | 10 - 11 | 1453 | 85 | ||
| AK168198.1 | - | 10 - 11 | 1483 | 85 | ||
| AK168369.1 | - | 10 - 11 | 1475 | 85 | ||
| AK168617.1 | - | 10 - 11 | 1473 | 85 | ||
| AK168879.1 | - | 10 - 11 | 1473 | 85 | ||
| AK168890.1 | - | 10 - 11 | 1473 | 85 | ||
| AK168932.1 | - | 10 - 11 | 1473 | 85 | ||
| AK169080.1 | - | 10 - 11 | 1476 | 85 | ||
| AK169084.1 | - | 10 - 11 | 1476 | 85 | ||
| AK169105.1 | - | 10 - 11 | 1473 | 85 | ||
| AK169142.1 | - | 10 - 11 | 1518 | 85 | ||
| AK169300.1 | - | 10 - 11 | 1473 | 85 | ||
| AK169308.1 | - | 10 - 11 | 1478 | 85 | ||
| AK169414.1 | - | 10 - 11 | 1476 | 85 | ||
| BC014854.1 | - | 10 - 11 | 1460 | 4500893 | 85 | |
| BC020417.1 | - | 10 - 11 | 1460 | 85 | ||
| BC055892.1 | - | 10 - 11 | 1910 | 85 | ||
| L01062.1 | - | 10 - 11 | 1460 | 85 | ||
More Information
Set Membership: |
|

Atp5a1
Genomic Map