Assay Details
| Gene Symbol: Atp6v1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007508.5 | NP_031534.2 | 7 - 8 | 946 | 90 | |
| GenBank mRNA | AK140873.1 | - | 7 - 8 | 922 | 90 | |
| AK149833.1 | - | 7 - 8 | 961 | 90 | ||
| AK150742.1 | - | 7 - 8 | 932 | 90 | ||
| AK153403.1 | - | 7 - 8 | 947 | 90 | ||
| AK154869.1 | - | 7 - 8 | 947 | 90 | ||
| AK160792.1 | - | 7 - 8 | 961 | 90 | ||
| AK166857.1 | - | 7 - 8 | 936 | 90 | ||
| AK170721.1 | - | 7 - 8 | 947 | 90 | ||
| BC038392.1 | - | 7 - 8 | 924 | 2651677 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1a
Genomic Map