Assay Details
Gene Symbol: Atp6v1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007508.5 | NP_031534.2 | 11 - 12 | 1365 | 78 | |
GenBank mRNA | AK140873.1 | - | 11 - 12 | 1341 | 78 | |
AK149833.1 | - | 11 - 12 | 1380 | 78 | ||
AK150742.1 | - | 11 - 12 | 1351 | 78 | ||
AK152785.1 | - | 1 - 2 | 79 | 78 | ||
AK153403.1 | - | 11 - 12 | 1366 | 78 | ||
AK154869.1 | - | 11 - 12 | 1366 | 78 | ||
AK160792.1 | - | 11 - 12 | 1380 | 78 | ||
AK166857.1 | - | 11 - 12 | 1355 | 78 | ||
AK170721.1 | - | 11 - 12 | 1366 | 78 | ||
BC038392.1 | - | 11 - 12 | 1343 | 2651677 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
