Assay Details
| Gene Symbol: Atp6v1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007509.3 | NP_031535.2 | 6 - 7 | 657 | 61 | |
| GenBank mRNA | AK146499.1 | - | 6 - 7 | 622 | 61 | |
| AK151200.1 | - | 6 - 7 | 658 | 61 | ||
| AK151322.1 | - | 6 - 7 | 630 | 61 | ||
| AK151586.1 | - | 6 - 7 | 630 | 61 | ||
| AK152718.1 | - | 6 - 7 | 630 | 61 | ||
| AK152766.1 | - | 6 - 7 | 630 | 61 | ||
| AK153341.1 | - | 6 - 7 | 621 | 61 | ||
| AK159133.1 | - | 6 - 7 | 628 | 61 | ||
| AK159153.1 | - | 6 - 7 | 628 | 61 | ||
| AK159586.1 | - | 6 - 7 | 625 | 61 | ||
| AK159701.1 | - | 6 - 7 | 629 | 61 | ||
| AK159986.1 | - | 6 - 7 | 629 | 61 | ||
| AK160080.1 | - | 6 - 7 | 628 | 61 | ||
| AK160854.1 | - | 6 - 7 | 623 | 61 | ||
| AK166669.1 | - | 6 - 7 | 597 | 61 | ||
| AK168852.1 | - | 6 - 7 | 621 | 61 | ||
| AK169155.1 | - | 6 - 7 | 636 | 61 | ||
| AK169270.1 | - | 6 - 7 | 633 | 61 | ||
| AK169420.1 | - | 6 - 7 | 625 | 61 | ||
| AK170110.1 | - | 6 - 7 | 653 | 61 | ||
| AK180494.1 | - | 1 - 2 | 61 | |||
| AK184499.1 | - | 2 - 3 | 209 | 61 | ||
| AK187885.1 | - | 1 - 2 | 76 | 61 | ||
| AK196120.1 | - | 1 - 2 | 76 | 61 | ||
| AK207339.1 | - | 1 - 2 | 76 | 61 | ||
| AK207413.1 | - | 1 - 2 | 76 | 61 | ||
| AK209086.1 | - | 1 - 2 | 76 | 61 | ||
| AK215006.1 | - | 1 - 2 | 76 | 61 | ||
| AK216515.1 | - | 3 - 4 | 265 | 61 | ||
| BC012497.1 | - | 6 - 7 | 622 | 4500843 | 61 | |
| BC046302.1 | - | 6 - 7 | 612 | 6492612 | 61 | |
| BC085300.1 | - | 6 - 7 | 604 | 6305977 | 61 | |
| Y12634.1 | - | 6 - 7 | 614 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1b2
Genomic Map