Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900001700.mm.1 |
NM_001285453.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001700.mm.1 |
TC1900000126.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900001701.mm.1 |
NM_001285453.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001701.mm.1 |
TC1900000126.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900001704.mm.1 |
NM_001285453.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001704.mm.1 |
NM_007522.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900001704.mm.1 |
TC1900000126.mm.1 |
MTA TC |