Assay Details
| Gene Symbol: Bid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007544.3 | NP_031570.2 | 4 - 5 | 506 | 76 | |
| GenBank mRNA | AK045731.1 | - | 4 - 5 | 521 | 76 | |
| AK051076.1 | - | 522 | 76 | |||
| AK052356.1 | - | 4 - 5 | 511 | 76 | ||
| AK077657.1 | - | 4 - 5 | 489 | 76 | ||
| AK161235.1 | - | 4 - 5 | 528 | 76 | ||
| BC002031.1 | - | 4 - 5 | 506 | 3486537 | 76 | |
| U75506.1 | - | 3 - 4 | 383 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bid
Genomic Map