Assay Details
Gene Symbol: C4bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007576.3 | NP_031602.3 | 7 - 8 | 1086 | 68 | |
GenBank mRNA | AB094485.1 | - | 1067 | 68 | ||
AB094486.1 | - | 7 - 8 | 1002 | 68 | ||
AK136735.1 | - | 7 - 8 | 1000 | 68 | ||
AK149468.1 | - | 7 - 8 | 1093 | 68 | ||
AK149477.1 | - | 7 - 8 | 1003 | 68 | ||
AK162345.1 | - | 7 - 8 | 998 | 68 | ||
BC012257.1 | - | 7 - 8 | 1058 | 4163203 | 68 | |
M17122.1 | - | 7 - 8 | 1100 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P60194 | M17122.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P60194 | BC012257.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P60194 | AK149477.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P60194 | AK162345.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P60194 | AK149468.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P60194 | M17122.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P60194 | BC012257.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P60194 | AK149477.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P60194 | AK162345.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P60194 | AK149468.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734187 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876277 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381186 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 679943 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376509 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764129 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 772432 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372782 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729597 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 429348 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 94038 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341445 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924856 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927743 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766172 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 177243 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68610 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108487 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171493 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P60194 | M17122.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P60194 | BC012257.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P60194 | AK149477.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P60194 | AK162345.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P60194 | AK149468.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P60194 | M17122.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P60194 | BC012257.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P60194 | AK149477.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P60194 | AK162345.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P60194 | AK149468.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20610897 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611030 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20610899 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611021 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611004 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611018 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611013 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611025 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611009 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611029 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611007 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611006 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611032 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611022 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20611015 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04262309 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171493 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04262300 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108487 | - | - |
Whole Mouse Genome G4122A | A_52_P293013 | AK050344.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P60194 | M17122.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P60194 | BC012257.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P60194 | AK149477.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P60194 | AK162345.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P60194 | AK149468.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P60194 | M17122.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P60194 | BC012257.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P60194 | AK149477.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P60194 | AK162345.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P60194 | AK149468.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106812 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108487 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171493 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00088905 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00088907 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00088906 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00088903 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00088908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20610883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20611054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20610910 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20610995 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01107709 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00957219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20610907 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20610909 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20610992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611019 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00285351 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611037 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20610998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611005 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611010 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611018 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20610911 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00957216 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611001 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01137895 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00810341 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611013 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611033 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00774908 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20610914 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20610994 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20611039 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10357519 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357524 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357518 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357528 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357526 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357523 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357517 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357527 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357520 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357522 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357521 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10357525 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418037_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10357516 | - | - |
Mouse Genome 430 2.0 Array | 1418037_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418037_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12269_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12269_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100024377.mm.1 | NM_007576.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100024377.mm.1 | NM_007576.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100024377.mm.1 | TC0100003041.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100024378.mm.1 | NM_007576.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100024378.mm.1 | NM_007576.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100024378.mm.1 | TC0100003041.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100024379.mm.1 | NM_007576.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100024379.mm.1 | NM_007576.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100024379.mm.1 | TC0100003041.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418037_PM_at | - | - |
Mouse Expression Set 430 | 1418037_at | - | - |
Murine Genome U74v2 Set | 102799_PM_at | - | - |
Murine Genome U74v2 Set | 102799_at | - | - |
Mouse Genome 430A 2.0 Array | 1418037_at | - | - |
Mouse Genome 430A 2.0 Array | 1418037_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418037_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418037_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 851543 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 105270484 | - | - |
MouseWG-6 v2 Expression BeadChip | 1690725 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 105270484 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1690725 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001690725 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2712176 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2712176 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1230710 | - | - |
mouse-6 v1.1 (Illumina) | scl16324.10_60-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 105270484 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1690725 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl16324.10_60-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105270484 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1690725 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2782794 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2782794 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230710 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2712176 | - | - |
