Assay Details
| Gene Symbol: C4bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007576.3 | NP_031602.3 | 8 - 9 | 1254 | 75 | |
| GenBank mRNA | AB094485.1 | - | 1235 | 75 | ||
| AB094486.1 | - | 8 - 9 | 1170 | 75 | ||
| AK136735.1 | - | 8 - 9 | 1168 | 75 | ||
| AK149468.1 | - | 8 - 9 | 1261 | 75 | ||
| AK149477.1 | - | 8 - 9 | 1171 | 75 | ||
| AK162345.1 | - | 8 - 9 | 1166 | 75 | ||
| BC012257.1 | - | 8 - 9 | 1226 | 4163203 | 75 | |
| M17122.1 | - | 8 - 9 | 1268 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

C4bp
Genomic Map