Assay Details
| Gene Symbol: Camk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174053.1 | NP_001167524.1 | 7 - 8 | 731 | 84 | |
| NM_001174054.1 | NP_001167525.1 | 7 - 8 | 731 | 84 | ||
| NM_007595.5 | NP_031621.3 | 7 - 8 | 731 | 84 | ||
| GenBank mRNA | AK046454.1 | - | 7 - 8 | 732 | 84 | |
| AK158794.1 | - | 4 - 5 | 492 | 84 | ||
| AK163698.1 | - | 7 - 8 | 548 | 84 | ||
| BC024803.1 | - | 7 - 8 | 547 | 84 | ||
| BC080273.1 | - | 7 - 8 | 533 | 6851995 | 84 | |
| X63615.1 | - | 7 - 8 | 546 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camk2b
Genomic Map