Assay Details
| Gene Symbol: Casp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042072.1 | - | 6 - 7 | 768 | 80 | |
| AK133829.1 | - | 6 - 7 | 779 | 80 | ||
| AK170649.1 | - | 5 - 6 | 501 | 80 | ||
| BC034262.1 | - | 6 - 7 | 834 | 4460089 | 80 | |
| D28492.1 | - | 6 - 7 | 757 | 80 | ||
| Y13085.1 | - | 6 - 7 | 836 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp2
Genomic Map