Assay Details
| Gene Symbol: Casp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042072.1 | - | 9 - 10 | 1137 | 80 | |
| AK133829.1 | - | 9 - 10 | 1148 | 80 | ||
| AK142569.1 | - | 2 - 3 | 276 | 80 | ||
| AK170649.1 | - | 8 - 9 | 870 | 80 | ||
| BC034262.1 | - | 9 - 10 | 1203 | 4460089 | 80 | |
| D28492.1 | - | 9 - 10 | 1126 | 80 | ||
| Y13085.1 | - | 9 - 10 | 1205 | 80 | ||
More Information
Set Membership: |
|

Casp2
Genomic Map