Assay Details
Gene Symbol: Cckbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF019371.1 | - | 3 - 4 | 650 | 68 | |
AK028325.1 | - | 3 - 4 | 860 | 68 | ||
AK039721.1 | - | 859 | 68 | |||
AK053307.1 | - | 3 - 4 | 720 | 68 | ||
BC103530.1 | - | 3 - 4 | 830 | 40039482 | 68 | |
BC103531.1 | - | 3 - 4 | 830 | 40039474 | 68 | |
BC103537.1 | - | 4 - 5 | 779 | 68 | ||
BC103538.1 | - | 3 - 4 | 830 | 40039475 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
