Assay Details
| Gene Symbol: Ccnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007630.2 | NP_031656.2 | 3 - 4 | 377 | 112 | |
| GenBank mRNA | AK076122.1 | - | 3 - 4 | 371 | 112 | |
| BC008247.1 | - | 3 - 4 | 293 | 3156950 | 112 | |
| CT010286.1 | - | 3 - 4 | 263 | 112 | ||
| X66032.1 | - | 3 - 4 | 364 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnb2
Genomic Map