Assay Details
| Gene Symbol: Ccnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007630.2 | NP_031656.2 | 4 - 5 | 548 | 99 | |
| GenBank mRNA | AK013312.1 | - | 4 - 5 | 517 | 99 | |
| AK076122.1 | - | 4 - 5 | 542 | 99 | ||
| BC008247.1 | - | 4 - 5 | 464 | 3156950 | 99 | |
| CT010286.1 | - | 4 - 5 | 434 | 99 | ||
| X66032.1 | - | 4 - 5 | 535 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnb2
Genomic Map