Assay Details
| Gene Symbol: Cd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159555.1 | NP_001153027.1 | 3 - 4 | 577 | 71 | |
| NM_001159556.1 | NP_001153028.1 | 3 - 4 | 437 | 71 | ||
| NM_001159557.1 | NP_001153029.1 | 4 - 5 | 488 | 71 | ||
| NM_001159558.1 | NP_001153030.1 | 4 - 5 | 604 | 71 | ||
| NM_007643.4 | NP_031669.3 | 3 - 4 | 580 | 71 | ||
| GenBank mRNA | AK128908.1 | - | 4 - 5 | 381 | 71 | |
| AK150648.1 | - | 3 - 4 | 401 | 71 | ||
| AK151355.1 | - | 3 - 4 | 416 | 71 | ||
| AK152904.1 | - | 1 - 2 | 87 | 71 | ||
| BC010262.1 | - | 3 - 4 | 389 | 3481681 | 71 | |
| L23108.1 | - | 3 - 4 | 398 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd36
Genomic Map