Assay Details
| Gene Symbol: Cd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159555.1 | NP_001153027.1 | 11 - 12 | 1583 | 121 | |
| NM_001159556.1 | NP_001153028.1 | 11 - 12 | 1443 | 121 | ||
| NM_001159557.1 | NP_001153029.1 | 12 - 13 | 1494 | 121 | ||
| NM_001159558.1 | NP_001153030.1 | 12 - 13 | 1610 | 121 | ||
| NM_007643.4 | NP_031669.3 | 11 - 12 | 1586 | 121 | ||
| GenBank mRNA | AK052825.1 | - | 11 - 12 | 1387 | 121 | |
| AK128908.1 | - | 12 - 13 | 1387 | 121 | ||
| AK151355.1 | - | 11 - 12 | 1422 | 121 | ||
| AK152904.1 | - | 9 - 10 | 1093 | 121 | ||
| AK153095.1 | - | 6 - 7 | 696 | 121 | ||
| AK172162.1 | - | 12 - 13 | 1432 | 121 | ||
| BC010262.1 | - | 11 - 12 | 1395 | 3481681 | 121 | |
| HQ171908.1 | - | 2 - 3 | 157 | 121 | ||
| L23108.1 | - | 11 - 12 | 1404 | 121 | ||
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as MTA transcript cluster(s) TC0500001992.mm.1
					            	This assay targets the same location as MTA transcript cluster(s) TC0500001992.mm.1| Set Membership: |  Ambion siRNA available  Amplicon length greater than or equal to 101  Apoptosis  Mammalian Gene Collection  Mus musculus  Probe spans exons | 
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Cd36
Cd36 Genomic Map
Genomic Map Back To Top
 Back To Top