Assay Details
| Gene Symbol: Cd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007650.3 | NP_031676.3 | 4 - 5 | 596 | 74 | |
| GenBank mRNA | AK042105.1 | - | 4 - 5 | 540 | 74 | |
| AK089076.1 | - | 4 - 5 | 545 | 74 | ||
| AK143736.1 | - | 4 - 5 | 572 | 74 | ||
| BC011258.1 | - | 4 - 5 | 596 | 4188683 | 74 | |
| M15177.1 | - | 4 - 5 | 474 | 74 | ||
| U65961.1 | - | 4 - 5 | 474 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd5
Genomic Map