Assay Details
| Gene Symbol: Cdk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007668.3 | NP_031694.1 | 1 - 2 | 85 | 93 | |
| GenBank mRNA | AK051790.1 | - | 1 - 2 | 85 | 93 | |
| BC052007.1 | - | 1 - 2 | 59 | 5706932 | 93 | |
| D29678.1 | - | 1 - 2 | 94 | 93 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK051790.1,BC052007.1,D29678.1,NM_007668.3 transcripts.
Set Membership: |
|

Cdk5
Genomic Map