Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600003538.mm.1 |
TC0600000444.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003538.mm.1 |
NM_001146351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003538.mm.1 |
NM_001146351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003538.mm.1 |
NM_007680.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003538.mm.1 |
NM_007680.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003539.mm.1 |
TC0600000444.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003539.mm.1 |
NM_001146351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003539.mm.1 |
NM_001146351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003539.mm.1 |
NM_007680.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003539.mm.1 |
NM_007680.4 |
Refseq |