Assay Details
Gene Symbol: Ephb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146351.1 | NP_001139823.1 | 15 - 16 | 3099 | 62 | |
NM_007680.4 | NP_031706.3 | 15 - 16 | 3095 | 62 | ||
GenBank mRNA | AK076442.1 | - | 15 - 16 | 2922 | 62 | |
AK087423.1 | - | 15 - 16 | 3010 | 62 | ||
AK163823.1 | - | 6 - 7 | 817 | 62 | ||
BC031924.1 | - | 14 - 15 | 2707 | 4911959 | 62 | |
L77867.1 | - | 15 - 16 | 2789 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
