Assay Details
| Gene Symbol: Chek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007691.5 | NP_031717.2 | 7 - 8 | 830 | 130 | |
| GenBank mRNA | AF016583.1 | - | 6 - 7 | 739 | 130 | |
| AK011258.1 | - | 7 - 8 | 774 | 130 | ||
| AK033179.1 | - | 7 - 8 | 800 | 130 | ||
| AK045336.1 | - | 7 - 8 | 772 | 130 | ||
| AK166675.1 | - | 7 - 8 | 831 | 130 | ||
| BC037613.1 | - | 6 - 7 | 543 | 130 | ||
| BC053394.1 | - | 7 - 8 | 767 | 131 | ||
| BC064445.1 | - | 6 - 7 | 1229 | 129 | ||
| BC100386.1 | - | 6 - 7 | 905 | 30481390 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chek1
Genomic Map