Assay Details
| Gene Symbol: Clcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007711.3 | NP_031737.1 | 7 - 8 | 1389 | 103 | |
| NM_173873.1 | NP_776298.1 | 8 - 9 | 1565 | 103 | ||
| NM_173874.1 | NP_776299.1 | 8 - 9 | 1565 | 103 | ||
| NM_173876.3 | NP_776301.1 | 7 - 8 | 1110 | 103 | ||
| GenBank mRNA | AF029347.1 | - | 7 - 8 | 1013 | 103 | |
| AK141136.1 | - | 8 - 9 | 1503 | 103 | ||
| AK147364.1 | - | 7 - 8 | 1545 | 103 | ||
| AK147548.1 | - | 7 - 8 | 1547 | 103 | ||
| AK147621.1 | - | 7 - 8 | 1547 | 103 | ||
| AK149276.1 | - | 8 - 9 | 1104 | 103 | ||
| AK150688.1 | - | 8 - 9 | 1494 | 103 | ||
| AK159698.1 | - | 8 - 9 | 1539 | 103 | ||
| AK161327.1 | - | 8 - 9 | 1493 | 103 | ||
| AK162639.1 | - | 8 - 9 | 1506 | 103 | ||
| AK169297.1 | - | 8 - 9 | 1486 | 103 | ||
| BC057133.1 | - | 7 - 8 | 1043 | 6833732 | 103 | |
| BC057855.1 | - | 7 - 8 | 1205 | 103 | ||
| CF536658.1 | - | 4 - 5 | 536 | 103 | ||
| X78874.1 | - | 7 - 8 | 1214 | 103 | ||
More Information
Set Membership: |
|

Clcn3
Genomic Map