Assay Details
| Gene Symbol: Ccr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007719.2 | NP_031745.2 | 1 - 2 | 91 | 104 | |
| GenBank mRNA | AK037965.1 | - | 1 - 2 | 92 | 104 | |
| AK154349.1 | - | 1 - 2 | 92 | 104 | ||
| AK154736.1 | - | 1 - 2 | 92 | 104 | ||
| AK154849.1 | - | 1 - 2 | 92 | 104 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK037965.1,AK154349.1,AK154736.1,AK154849.1,NM_007719.2 transcripts.
Set Membership: |
|

Ccr7
Genomic Map