Assay Details
| Gene Symbol: Cox5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007747.2 | NP_031773.2 | 1 - 2 | 186 | 115 | |
| GenBank mRNA | AK018723.1 | - | 1 - 2 | 121 | 115 | |
| BC003831.1 | - | 1 - 2 | 144 | 115 | ||
| BC034302.1 | - | 1 - 2 | 139 | 5372849 | 115 | |
| BY259203.1 | - | 1 - 2 | 187 | 115 | ||
| CO044753.1 | - | 4 - 5 | 115 | |||
| X15963.1 | - | 1 - 2 | 110 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542211 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542218 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542220 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542216 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542219 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542214 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542217 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542212 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542213 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P335900 | X15963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P335900 | X15963.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 553765 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764859 | NM_146503.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764859 | NM_207141.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936428 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437442 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347273 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145466 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64292 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118383 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141798 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P335900 | X15963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P335900 | X15963.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P335900 | X15963.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P335900 | X15963.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542226 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118383 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06801898 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141798 | - | - |
| Whole Mouse Genome G4122A | A_51_P335900 | X15963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P335900 | X15963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121110 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121497 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141798 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118383 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07495289 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08174155 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08174151 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08174141 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542211 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542216 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542214 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542219 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542212 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542217 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542218 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08155022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08174140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07771177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08032972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07698837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07597222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08174151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08174142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08184409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07956983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542243 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10585711 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585707 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585708 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585710 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585712 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585709 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415933_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439267_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448153_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10585706 | - | - |
| Mouse Genome 430 2.0 Array | 1439267_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1448153_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415933_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448153_at | - | - |
| Mouse Genome 430 2.0 Array | 1439267_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1415933_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12858_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12858_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900005181.mm.1 | NM_007747.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900005181.mm.1 | NM_007747.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900005181.mm.1 | TC0900000705.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900005183.mm.1 | NM_007747.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900005183.mm.1 | NM_007747.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900005183.mm.1 | TC0900000705.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900005180.mm.1 | NM_007747.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900005180.mm.1 | NM_007747.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900005180.mm.1 | TC0900000705.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900005182.mm.1 | NM_007747.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900005182.mm.1 | NM_007747.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900005182.mm.1 | TC0900000705.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439267_PM_x_at | - | - |
| Mouse Expression Set 430 | 1448153_at | - | - |
| Mouse Expression Set 430 | 1415933_PM_a_at | - | - |
| Mouse Expression Set 430 | 1439267_x_at | - | - |
| Mouse Expression Set 430 | 1415933_a_at | - | - |
| Mouse Expression Set 430 | 1448153_PM_at | - | - |
| Murine Genome U74v2 Set | 161490_at | - | - |
| Murine Genome U74v2 Set | 160438_at | - | - |
| Murine Genome U74v2 Set | 161490_PM_at | - | - |
| Murine Genome U74v2 Set | 160438_PM_at | - | - |
| Murine Genome U74v2 Set | 171593_at | - | - |
| Murine Genome U74v2 Set | 171593_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415933_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448153_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448153_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415933_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439267_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439267_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415933_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439267_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415933_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439267_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448153_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448153_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1450609 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1450609 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450609 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2682800 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2682800 | - | - |
| mouse-6 v1.1 (Illumina) | scl36904.3.10_19-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450609 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36904.3.10_19-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450609 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2682800 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2682800 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36904.3.10_19-S | - | - |

Cox5a
Genomic Map